Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Valerylglycine Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022385 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2385 250 mg
11-102-2384 10 mg
11-102-2386 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-102-2385 Supplier MilliporeSigma Supelco Supplier No. 04268250MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

CAS 24003-66-5
Molecular Formula C7H13NO3
Linear Formula C7H13NO3
Quantity 250 mg
MDL Number MFCD01910091
UN Number NONH for all modes of transport
Synonym 2-(Pentanoylamino)acetic acid; N-(1-Oxopentyl)glycine; N-Pentanoylglycine; Valeryl glycine
Formula Weight 159.18
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.