Learn More
N-Valerylglycine Analytical Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 24003-66-5 |
| Molecular Formula | C7H13NO3 |
| Linear Formula | C7H13NO3 |
| MDL Number | MFCD01910091 |
| Quantity | 50 mg |
| UN Number | NONH for all modes of transport |
| Synonym | 2-(Pentanoylamino)acetic acid; N-(1-Oxopentyl)glycine; N-Pentanoylglycine; Valeryl glycine |
| Formula Weight | 159.18 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.