Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-valerylglycine-d2 | 3008541-76-9 | 95.0% | 161.20 g·mol⁻¹ | C7H11D2NO3 | 1 MG
SDP

Catalog No. 5000283054
Encompass_Preferred

A deuterium-labeled form of N-valerylglycine supplied as a 1 mg research standard for use as a stable isotope internal standard or tracer in analytical and metabolite studies. It supports improved quantitation and tracing in mass spectrometry workflows and is intended for research applications with appropriate storage to maintain stability.

  • Stable isotope-labeled internal standard for mass spectrometry.
  • Supplied as a 1 mg research-grade quantity.
  • Purity 95.0%.
  • Molecular formula C7H11D2NO3; molecular weight 161.20 g·mol⁻¹.
  • Suitable for analytical method development and tracer studies.
  • Storage recommended at low temperatures to preserve stability.

Catalog No. 50-002-83054 Supplier Medchemexpress LLC Supplier No. HYW141934S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A deuterium-labeled form of N-valerylglycine supplied as a 1 mg research standard for use as a stable isotope internal standard or tracer in analytical and metabolite studies. It supports improved quantitation and tracing in mass spectrometry workflows and is intended for research applications with appropriate storage to maintain stability.

  • Stable isotope-labeled internal standard for mass spectrometry.
  • Supplied as a 1 mg research-grade quantity.
  • Purity 95.0%.
  • Molecular formula C7H11D2NO3; molecular weight 161.20 g·mol⁻¹.
  • Suitable for analytical method development and tracer studies.
  • Storage recommended at low temperatures to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.