Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-valerylglycine-d2 | 3008541-76-9 | 95.0% | 161.20 g/mol | C7H11D2NO3 | 10 MG
SDP

Catalog No. 5000283055
Encompass_Preferred

Deuterated N-valerylglycine-d2 is a stable isotope-labeled amino acid derivative intended for use as an internal standard or tracer in analytical laboratories. It is provided as a small, high-purity material for quantitative analysis and method validation in spectroscopy and mass spectrometry workflows.

  • Deuterium-labeled tracer for quantitative NMR, GC-MS, and LC-MS.
  • High purity: 95.0%.
  • Molecular formula C7H11D2NO3 and molecular weight 161.20 g/mol.
  • Supplied as 10 MG small-scale analytical quantity.
  • Stable when stored under recommended conditions, supporting reproducible results.
  • Suitable as an internal standard for method validation and accurate quantitation.

Catalog No. 50-002-83055 Supplier Medchemexpress LLC Supplier No. HYW141934S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Deuterated N-valerylglycine-d2 is a stable isotope-labeled amino acid derivative intended for use as an internal standard or tracer in analytical laboratories. It is provided as a small, high-purity material for quantitative analysis and method validation in spectroscopy and mass spectrometry workflows.

  • Deuterium-labeled tracer for quantitative NMR, GC-MS, and LC-MS.
  • High purity: 95.0%.
  • Molecular formula C7H11D2NO3 and molecular weight 161.20 g/mol.
  • Supplied as 10 MG small-scale analytical quantity.
  • Stable when stored under recommended conditions, supporting reproducible results.
  • Suitable as an internal standard for method validation and accurate quantitation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.