Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-valerylglycine-d2 | 3008541-76-9 | 95.0% | 161.20 g/mol | C7H11D2NO3 | 5 MG
SDP

Catalog No. 5000283800
Encompass_Preferred

N-Valerylglycine-d2 is the deuterium-labeled (d2) analogue of N-valerylglycine provided for laboratory research use. It is designed as a stable-isotope tracer or internal standard to support accurate quantitation in analytical workflows such as NMR, GC-MS, and LC-MS. Supplied as a solid with defined purity and identified by its CAS number for unambiguous reference.

  • Deuterium-labeled analogue suitable as tracer or internal standard.
  • Supports quantitative analysis by NMR, GC-MS, and LC-MS.
  • Molecular formula C7H11D2NO3 and molecular weight 161.20 g/mol.
  • CAS number 3008541-76-9 for unambiguous identification.
  • Available in laboratory pack sizes, including 5 MG.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-83800 Supplier Medchemexpress LLC Supplier No. HYW141934S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Valerylglycine-d2 is the deuterium-labeled (d2) analogue of N-valerylglycine provided for laboratory research use. It is designed as a stable-isotope tracer or internal standard to support accurate quantitation in analytical workflows such as NMR, GC-MS, and LC-MS. Supplied as a solid with defined purity and identified by its CAS number for unambiguous reference.

  • Deuterium-labeled analogue suitable as tracer or internal standard.
  • Supports quantitative analysis by NMR, GC-MS, and LC-MS.
  • Molecular formula C7H11D2NO3 and molecular weight 161.20 g/mol.
  • CAS number 3008541-76-9 for unambiguous identification.
  • Available in laboratory pack sizes, including 5 MG.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.