Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Vanillyldecanamide | 31078-36-1 | MFCD01233545 | 50 MG
SDP

Catalog No. 5000418570
Encompass_Preferred

N-Vanillyldecanamide is a capsaicinoid isolated from the fruits of Capsicum annuum. It significantly reduced the radical length of Lactuca sativa seedling in a dose-dependent manner. This product is for research use only.

  • Purity of 99.86%
  • Molecular weight of 307.43
  • Molecular formula C18H29NO3
  • Appears as a white to off-white solid
  • Exhibits agonist activity at rat TRPV1 in CHO cells with an EC50 of 83 nM

Catalog No. 50-004-18570 Supplier Medchemexpress LLC Supplier No. HYN509950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Vanillyldecanamide is a capsaicinoid isolated from the fruits of Capsicum annuum. It significantly reduced the radical length of Lactuca sativa seedling in a dose-dependent manner. This product is for research use only.

  • Purity of 99.86%
  • Molecular weight of 307.43
  • Molecular formula C18H29NO3
  • Appears as a white to off-white solid
  • Exhibits agonist activity at rat TRPV1 in CHO cells with an EC50 of 83 nM

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.