Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-vanillyldecanamide | 31078-36-1 | MFCD01233545 | 99.9% | 307.43 g·mol⁻¹ | C18H29NO3 | 10 MG
SDP

Catalog No. 5000289839
Encompass_Preferred

N-Vanillyldecanamide is a capsaicinoid research chemical (CAS 31078-36-1) isolated from the fruits of Capsicum annuum. It has been reported to reduce the radical length of Lactuca sativa seedlings in a dose-dependent manner. The compound is supplied for laboratory research use with supporting documentation for handling and analysis.

  • High purity (99.86%) suitable for analytical and biological studies.
  • Molecular formula C18H29NO3 and molecular weight 307.43 g·mol⁻¹.
  • Supplied in small laboratory quantities, including a 10 mg pack size.
  • Available as a solid and as 10 mM solutions in DMSO for convenience.
  • Storage: keep at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Typical applications include capsaicinoid activity studies, plant physiology research, and structure-activity investigations.
  • Product documentation includes datasheet, COA, and SDS for quality and safety reference.

Catalog No. 50-002-89839 Supplier Medchemexpress LLC Supplier No. HYN509910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Vanillyldecanamide is a capsaicinoid research chemical (CAS 31078-36-1) isolated from the fruits of Capsicum annuum. It has been reported to reduce the radical length of Lactuca sativa seedlings in a dose-dependent manner. The compound is supplied for laboratory research use with supporting documentation for handling and analysis.

  • High purity (99.86%) suitable for analytical and biological studies.
  • Molecular formula C18H29NO3 and molecular weight 307.43 g·mol⁻¹.
  • Supplied in small laboratory quantities, including a 10 mg pack size.
  • Available as a solid and as 10 mM solutions in DMSO for convenience.
  • Storage: keep at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Typical applications include capsaicinoid activity studies, plant physiology research, and structure-activity investigations.
  • Product documentation includes datasheet, COA, and SDS for quality and safety reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.