Learn More
Selleck Chemical LLC N-Vanillyldecanamide
SDP
Catalog No. 5000002401
Encompass_Preferred
N-Vanillyldecanamide is a capsaicinoid isolated from the fruits of Capsicum annuum which can significantly reduce the radical length of Lactuca sativa seedling in a dose-dependent manner
Catalog No. 50-000-02401 Supplier Selleck Chemical LLC Supplier No. S69755MG
N-Vanillyldecanamide is a capsaicinoid isolated from the fruits of Capsicum annuum which can significantly reduce the radical length of Lactuca sativa seedling in a dose-dependent manner