Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Vanillylnonanamide, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019408 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9408 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9408 Supplier MilliporeSigma Supelco Supplier No. 1597150MG
Add to Cart
Add to Cart

N-Vanillylnonanamide has antifouling properties. It acts as an nicotinamide adenine dinucleotide phosphate (NADPH) inhibitor.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 2444-46-4
Flash Point 190°C
Physical Form Neat
Linear Formula C17H27 NO3
MDL Number MFCD00017286
Synonym Nonivamide; Capsaicin synthetic; N-(4-Hydroxy-3-methoxybenzyl)nonanamide; N-Vanillylpelargonamide; Nonanoic acid vanillylamide; Nonylvanylamide; Pelargonic acid vanillylamide; Pseudocapsaicin
Type Analytical Standard
Quantity 50 mg
Formula Weight 293.40
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.