Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Vanillylnonanamide 98.0+%, TCI America™
SDP

Catalog No. M090010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M090010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M090010G Supplier TCI America Supplier No. M090010G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2444-46-4
Molecular Formula C17H27NO3
Molecular Weight (g/mol) 293.407
MDL Number MFCD00017286
InChI Key RGOVYLWUIBMPGK-UHFFFAOYSA-N
Synonym N-Nonylvanylamide, N-Pelargonic Acid Vanillylamide, N-Pelargonylvanillylamide, Capsaicin (Synthetic)
PubChem CID 2998
ChEBI CHEBI:46936
IUPAC Name N-[(4-hydroxy-3-methoxyphenyl)methyl]nonanamide
SMILES CCCCCCCCC(=O)NCC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 54°C
Quantity 10 g
UN Number 2811
Formula Weight 293.41
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Lumps
Chemical Name or Material N-Vanillylnonanamide [=Capsaicin (Synthetic)]
EINECSNumber (9)-0993
RTECSNumber RA5955000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.