Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Vinylacetamide 98.0+%, TCI America™
SDP

Catalog No. V01055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01055G 5 g
V010525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01055G Supplier TCI America Supplier No. V01055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5202-78-8
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00081205
InChI Key RQAKESSLMFZVMC-UHFFFAOYSA-N
PubChem CID 78875
IUPAC Name N-ethenylacetamide
SMILES CC(=O)NC=C

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 85.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Vinylacetamide
EINECSNumber (2)-3701
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.