Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Vinylcarbazole, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1484-13-5
C14H11N
193.24
MFCD00004966
KKFHAJHLJHVUDM-UHFFFAOYSA-N
9-vinylcarbazole,n-vinylcarbazole,9-vinyl-9h-carbazole,9h-carbazole, 9-ethenyl,n-vinyl carbazole,vinylcarbazole,1-vinylcarbazole,poly 9-vinylcarbazole,carbazole, 9-vinyl,poly-n-vinylcarbazole
15143
9-ethenylcarbazole
C=CN1C2=CC=CC=C2C3=CC=CC=C31
1484-13-5
C14H11N
193.24
MFCD00004966
KKFHAJHLJHVUDM-UHFFFAOYSA-N
9-vinylcarbazole,n-vinylcarbazole,9-vinyl-9h-carbazole,9h-carbazole, 9-ethenyl,n-vinyl carbazole,vinylcarbazole,1-vinylcarbazole,poly 9-vinylcarbazole,carbazole, 9-vinyl,poly-n-vinylcarbazole
15143
9-ethenylcarbazole
C=CN1C2=CC=CC=C2C3=CC=CC=C31
Boiling Point 154°C (3.0mmHg)
Flash Point 182°C
Melting Point 63°C to 65°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 193.24
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material N-Vinylcarbazole
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-055-