Learn More
AdipoGen N-Z-L-Homoserine lactone
SDP
Catalog No. 502560687
Encompass_Preferred
Chemical. CAS 35677-89-5. Formula C12H13NO4. MW 235.24. N-Z-L-Homoserine lactone is a synthetic building block or intermediate used in the synthesis of biological active agents and analogs of bacterial quorum-sensing signaling molecules. N-Z-L-Homoserine lactone is shown to irreversibly inactivate the 3C cysteine proteinase of hepatitis A virus HAV and might be a scaffold for potent cyteine protein inhibtiors. Shown to reduce the lag phase duration LPD and increase the exponential growth rate EGR of three strains of Pseudomonas fluorescens isolated from refrigerated raw milk.
Catalog No. 50-256-0687 Supplier AdipoGen Supplier No. CDXH0379G005
Chemical. CAS 35677-89-5. Formula C12H13NO4. MW 235.24. N-Z-L-Homoserine lactone is a synthetic building block or intermediate used in the synthesis of biological active agents and analogs of bacterial quorum-sensing signaling molecules. N-Z-L-Homoserine lactone is shown to irreversibly inactivate the 3C cysteine proteinase of hepatitis A virus HAV and might be a scaffold for potent cyteine protein inhibtiors. Shown to reduce the lag phase duration LPD and increase the exponential growth rate EGR of three strains of Pseudomonas fluorescens isolated from refrigerated raw milk.