Learn More
Cayman Chemical N-[1-(2,3-Di(9-octadecenoyloxy)propyl]-N,N,N-trimethylammonium (chloride), Purity: ≥98% (mixture of isomers), CAS: 104872-42-6, 50 mg
SDP
Catalog No. 502919083
Encompass_Preferred
A mixture of the cationic lipid isomers DOTMA and N-[1-(2,3-dielaidoyloxy)propyl]-N,N,N-trimethylammonium
Catalog No. 50-291-9083 Supplier Cayman Chemical Supplier No. 3859250 MG
A mixture of the cationic lipid isomers DOTMA and N-[1-(2,3-dielaidoyloxy)propyl]-N,N,N-trimethylammonium