Learn More
Cayman Chemical DOTMA (chloride), Purity: ≥95%, CAS: 104162-48-3, 50 mg
SDP
Catalog No. 502933361
Encompass_Preferred
A cationic lipid; has been used as a component in liposomes that can be used to encapsulate siRNA, microRNAs, and oligonucleotides and for gene transfection in vitro
Catalog No. 50-293-3361 Supplier Cayman Chemical Supplier No. 2592650 MG
A cationic lipid; has been used as a component in liposomes that can be used to encapsulate siRNA, microRNAs, and oligonucleotides and for gene transfection in vitro