Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N1-(3-Trimethoxysilylpropyl)Diethylenetriamine

Catalog No. 413348100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-334-8100ML 100 mL
41-334-8500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 41-334-8100ML Supplier Sigma Aldrich Supplier No. 413348100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N1-(3-Trimethoxysilylpropyl)diethylenetriamine (TRI-silane) can be used as a grafting agent to incorporate amine functional group onto the surface of various ordered mesoporous silica. It can be also used in CO2 capture applications.

Specifications

CAS 35141-30-1
Density 1.03 g/mL at 25°C
Boiling Point 114°C to 118°C (2 mmHg, lit.)
Molecular Formula C10H27N3O3Si
Refractive Index n20/D 1.459
Linear Formula (CH3O)3Si(CH2)3NHCH2CH2NHCH2CH2NH2
MDL Number MFCD00069173
Quantity 100 mL
Synonym 2-[2-(3-Trimethoxysilylpropylamino)ethylamino]ethylamine; 3-[2-(2-Aminoethylamino)ethylamino]propyltrimethoxysilane
Molecular Weight (g/mol) 265.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.