Learn More
Medchemexpress LLC N1-(4-methylphenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide | 144217-65-2 | 99.0% | 317.43 | C18H27N3O2 | 100 MG
SDP
Catalog No. 5000215005
Encompass_Preferred
YYA-021 is a small-molecule CD4 mimic that inhibits HIV entry, exhibiting high anti-HIV activity and low cytotoxicity. It is provided for research use to study HIV entry mechanisms, antiviral screening, and cell-based assay development.
  • Small-molecule CD4 mimic that targets gp120-mediated entry.
  • Inhibits HIV entry with demonstrated antiviral activity.
  • Low cytotoxicity suitable for cell-based assays.
  • Well-characterized chemical properties: C18H27N3O2; molecular weight 317.43.
  • High purity for research applications (>98.9%).
  • Stable solid; recommended storage at -20°C (powder) and -80°C (in solvent).
  • Supplied in milligram quantities convenient for assay development.
Catalog No. 50-002-15005 Supplier Medchemexpress LLC Supplier No. HY100039100MG
YYA-021 is a small-molecule CD4 mimic that inhibits HIV entry, exhibiting high anti-HIV activity and low cytotoxicity. It is provided for research use to study HIV entry mechanisms, antiviral screening, and cell-based assay development.
  • Small-molecule CD4 mimic that targets gp120-mediated entry.
  • Inhibits HIV entry with demonstrated antiviral activity.
  • Low cytotoxicity suitable for cell-based assays.
  • Well-characterized chemical properties: C18H27N3O2; molecular weight 317.43.
  • High purity for research applications (>98.9%).
  • Stable solid; recommended storage at -20°C (powder) and -80°C (in solvent).
  • Supplied in milligram quantities convenient for assay development.