Learn More
Medchemexpress LLC N1-(5-chloro-4-(5-(cyclopropylmethyl)-1-methyl-1H-pyrazol-4-yl)pyrimidin-2-yl)cyclohexane-1,4-diamine | 2079068-74-7 | 98.6% | 360.88 g·mol⁻¹ | C18H25ClN6 | 10MG
SDP
Catalog No. 5000235561
Encompass_Preferred
BTX-A51 is a potent, orally active small-molecule inhibitor of casein kinase 1α (CK1α) with reported anti-leukemic activity; it induces leukemia cell apoptosis and is used as a research reagent in biochemical and cellular studies.
  • Potent inhibitor of casein kinase 1α (CK1α).
  • Orally active small-molecule suitable for in vivo and in vitro studies.
  • Reported to induce leukemia cell apoptosis and demonstrate anti-leukemic activity.
  • High purity, typically 98.58% as supplied.
  • Solid form, white to light yellow appearance.
  • Defined chemical identity: C18H25ClN6, MW 360.88 g·mol⁻¹.
  • Available in small research pack sizes including 10 mg.
Catalog No. 50-002-35561 Supplier Medchemexpress LLC Supplier No. HY12395410MG
Add to Cart
Add to Cart
BTX-A51 is a potent, orally active small-molecule inhibitor of casein kinase 1α (CK1α) with reported anti-leukemic activity; it induces leukemia cell apoptosis and is used as a research reagent in biochemical and cellular studies.
  • Potent inhibitor of casein kinase 1α (CK1α).
  • Orally active small-molecule suitable for in vivo and in vitro studies.
  • Reported to induce leukemia cell apoptosis and demonstrate anti-leukemic activity.
  • High purity, typically 98.58% as supplied.
  • Solid form, white to light yellow appearance.
  • Defined chemical identity: C18H25ClN6, MW 360.88 g·mol⁻¹.
  • Available in small research pack sizes including 10 mg.