Learn More
Medchemexpress LLC N1-Acetylspermidine hydrochloride | 34450-16-3 | 99.1% | 260.20 | 10 MG
SDP
Catalog No. 5000436144
Encompass_Preferred
N1-Acetylspermidine hydrochloride is an acetyl derivative of polyamines and acts as a substrate for polyamine oxidase (PAO). It is utilized in colorectal cancer research and can promote apoptosis when combined with Procyanidins.
  • Acetyl derivative of polyamines
  • Acts as a substrate for polyamine oxidase (PAO)
  • Promotes apoptosis when combined with Procyanidins
  • Exhibits cleavage efficiency at apurinic sites of DNA
  • Useful in colorectal cancer research
Catalog No. 50-004-36144 Supplier Medchemexpress LLC Supplier No. HY113056A10MG
N1-Acetylspermidine hydrochloride is an acetyl derivative of polyamines and acts as a substrate for polyamine oxidase (PAO). It is utilized in colorectal cancer research and can promote apoptosis when combined with Procyanidins.
  • Acetyl derivative of polyamines
  • Acts as a substrate for polyamine oxidase (PAO)
  • Promotes apoptosis when combined with Procyanidins
  • Exhibits cleavage efficiency at apurinic sites of DNA
  • Useful in colorectal cancer research