Learn More
Cayman Chemical N1-Acetylspermidine (hydrochloride), 34450-16-3, 1 mg
SDP
Supplier:  Cayman Chemical 90015351
Encompass_Preferred
Molecular Formula C9H21N3O • 2HCl, Purity ≥98%, Formula Weight 260.2, Synonyms: N1-AcSPD; A substrate for polyamine oxidase that is oxidized to 3-acetamidopropanal and putrescine, a polyamine involved in the division, differentiation, and membrane function of cells; used to examine the interaction between DNA and polyamines during the cleavage of phosphodiester bonds at apurinic/apyrimidinic sites in DNA.
Catalog No. NC1062693