Learn More
Medchemexpress LLC N1-acetylspermine-d3 trihydrochloride | 77928-70-2 | MFCD00083219 | 99.9% | 356.78 g/mol | C12H28D3Cl3N4O | 1 MG
SDP
Catalog No. 5000284363
Encompass_Preferred
N1-acetylspermine-d3 trihydrochloride is a deuterium-labeled isotopic standard of N1-acetylspermine provided as the trihydrochloride salt. It is intended for use as an internal standard in analytical methods, particularly mass spectrometry and metabolomics studies of polyamine metabolism.
  • Deuterium-labeled internal standard suitable for mass spectrometry quantification.
  • High isotopic and chemical purity for accurate tracing and minimal interference.
  • Trihydrochloride salt form improves stability and solubility in aqueous solutions.
  • Supplied in small analytical pack sizes appropriate for method development.
Catalog No. 50-002-84363 Supplier Medchemexpress LLC Supplier No. HYW6540401MG
N1-acetylspermine-d3 trihydrochloride is a deuterium-labeled isotopic standard of N1-acetylspermine provided as the trihydrochloride salt. It is intended for use as an internal standard in analytical methods, particularly mass spectrometry and metabolomics studies of polyamine metabolism.
  • Deuterium-labeled internal standard suitable for mass spectrometry quantification.
  • High isotopic and chemical purity for accurate tracing and minimal interference.
  • Trihydrochloride salt form improves stability and solubility in aqueous solutions.
  • Supplied in small analytical pack sizes appropriate for method development.