Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N1-Ethyl-4-(trifluoromethoxy)aniline, Technical Grade, Thermo Scientific™

Catalog No. TL00676EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL00676EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00676EA Supplier Thermo Scientific Chemicals Supplier No. TL00676EA
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 175278-20-3
Packaging Amber Glass Bottle
Molecular Formula C9H10F3NO
Linear Formula CF3OC6H4NHCH2CH3
Quantity 10 g
Formula Weight 205.18
Percent Purity Tech.
Grade Technical
Chemical Name or Material N1-Ethyl-4-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.