Learn More
Bachem [H-Glu(Cys-Gly-[3-(4-amino-butylamino)-propyl]-amide)-OH]2
SDP
Supplier:  Bachem 4015761.0025
Encompass_Preferred
N1-Glutathionyl-spermidine disulfide, [H-Glu(Cys-Gly-[3-(4-amino-butylamino)-propyl]-amide)-OH]2, 108081-77-2, C34H66N12O10S2, Mr 867.10, 25mg. The enzyme trypanothione reductase from the insect trypanosomatid Crithidia fasciculata not only uses trypanothione as a substrate, but also this related acyclic disulfide (Km = 149 uM).
Catalog No. 50-259-465