Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N1-guanyl-1,7-diaminoheptane sulfate | 150417-90-6 | 98.0% | 270.35 g·mol⁻¹ | C8H22N4O4S | 5 MG
SDP

Catalog No. 5000243331
Encompass_Preferred

GC7 sulfate is the sulfate salt of N1-guanyl-1,7-diaminoheptane, a selective deoxyhypusine synthase (DHPS) inhibitor used as a research reagent to study hypusination, eukaryotic translation, and related cellular processes.

  • Selective deoxyhypusine synthase inhibitor for research use.
  • Sulfate salt supplied as a solid.
  • High purity (98.0%).
  • Molecular weight 270.35 g·mol⁻¹.
  • Molecular formula C8H22N4O4S.
  • Available in small research quantities (for example, 5 mg).

Catalog No. 50-002-43331 Supplier Medchemexpress LLC Supplier No. HY108314A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GC7 sulfate is the sulfate salt of N1-guanyl-1,7-diaminoheptane, a selective deoxyhypusine synthase (DHPS) inhibitor used as a research reagent to study hypusination, eukaryotic translation, and related cellular processes.

  • Selective deoxyhypusine synthase inhibitor for research use.
  • Sulfate salt supplied as a solid.
  • High purity (98.0%).
  • Molecular weight 270.35 g·mol⁻¹.
  • Molecular formula C8H22N4O4S.
  • Available in small research quantities (for example, 5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.