Learn More
Apexbio Technology LLC N1-Methylguanosine-5'-Triphosphate 50ul (100 mM)
SDP
Catalog No. 5000063846
Encompass_Preferred
N1-Methylguanosine-5 -Triphosphate (N1-meGTP) is a modified nucleotide analog featuring methylation at the N1 position of guanine Upon incorporation into RNA transcripts during in vitro transcription experiments the N1-methyl modification influences RNA structure binding affinity with RNA-interacting proteins translational regulation and RNA stability Such methylated RNA transcripts serve as research tools for investigating the biological roles of RNA methylation in gene expression modulation especially regarding translation initiation mechanisms and RNA-protein interactions Additionally N1-meGTP-based RNA modification is relevant in studies exploring RNA methylation alterations observed in viral infections and cancer pathogenesis offering potential implications for antiviral and oncology therapeutic insights
Catalog No. 50-000-63846 Supplier Apexbio Technology LLC Supplier No. B79910.05
N1-Methylguanosine-5 -Triphosphate (N1-meGTP) is a modified nucleotide analog featuring methylation at the N1 position of guanine Upon incorporation into RNA transcripts during in vitro transcription experiments the N1-methyl modification influences RNA structure binding affinity with RNA-interacting proteins translational regulation and RNA stability Such methylated RNA transcripts serve as research tools for investigating the biological roles of RNA methylation in gene expression modulation especially regarding translation initiation mechanisms and RNA-protein interactions Additionally N1-meGTP-based RNA modification is relevant in studies exploring RNA methylation alterations observed in viral infections and cancer pathogenesis offering potential implications for antiviral and oncology therapeutic insights