Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N1-Methyl-3-chloroaniline, 95%, Thermo Scientific™

Catalog No. TL00250EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL00250EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00250EA Supplier Thermo Scientific Chemicals Supplier No. TL00250EA
Add to Cart

Chemical Identifiers

CAS 7006-52-2
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00052014
InChI Key WFGYSQDPURFIFL-UHFFFAOYSA-N
Synonym n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline
PubChem CID 138900
IUPAC Name 3-chloro-N-methylaniline
SMILES CNC1=CC=CC(Cl)=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula ClC6H4NHCH3
Quantity 10 g
Formula Weight 141.6
Percent Purity 95%
Chemical Name or Material N1-methyl-3-chloroaniline
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.