Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N1-Methyl-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™

Catalog No. TL00405EB
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL00405EB 25 g
TL00405EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TL00405EB Supplier Thermo Scientific Chemicals Supplier No. TL00405EB
Add to Cart

Chemical Identifiers

CAS 41419-59-4
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00052332
InChI Key MGCCWCLGIPNIBP-UHFFFAOYSA-N
Synonym n-methyl-4-trifluoromethoxy aniline, n1-methyl-4-trifluoromethoxy aniline, benzenamine,n-methyl-4-trifluoromethoxy, methyl 4-trifluoromethoxy phenyl amine, pubchem8511, 4-trifluoromethoxy-n-methylaniline, methyl 4-trifluoromethoxyphenyl amine, methyl-4-trifluoromethoxy-phenyl-amine, n-methyl-4-trifluoromethoxyphenyl amine
PubChem CID 737351
IUPAC Name N-methyl-4-(trifluoromethoxy)aniline
SMILES CNC1=CC=C(C=C1)OC(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Linear Formula CF3OC6H4NHCH3
Quantity 25 g
Formula Weight 191.15
Percent Purity 95%
Chemical Name or Material N1-methyl-4-(trifluoromethoxy)aniline
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.