Learn More
Apexbio Technology LLC N1-Methylpseudo-UTP (D3) 50ul (100 mM)
SDP
Catalog No. 5000089317
Encompass_Preferred
N1-Methylpseudo-UTP D3 is a chemically modified nucleotide analog labeled with deuterium It is designed to substitute conventional uridine triphosphate during enzymatic transcription reactions allowing for the synthesis of isotope-labeled RNA N1-Methylpseudo-UTP D3 exerts its function through incorporation into RNA thereby conferring distinct structural and biochemical properties to RNA transcripts Based on these properties N1-Methylpseudo-UTP D3 holds research potential in structural analyses of RNA RNA-protein interaction studies and biophysical investigations including nuclear magnetic resonance (NMR) spectroscopy
Catalog No. 50-000-89317 Supplier Apexbio Technology LLC Supplier No. B88170.05
N1-Methylpseudo-UTP D3 is a chemically modified nucleotide analog labeled with deuterium It is designed to substitute conventional uridine triphosphate during enzymatic transcription reactions allowing for the synthesis of isotope-labeled RNA N1-Methylpseudo-UTP D3 exerts its function through incorporation into RNA thereby conferring distinct structural and biochemical properties to RNA transcripts Based on these properties N1-Methylpseudo-UTP D3 holds research potential in structural analyses of RNA RNA-protein interaction studies and biophysical investigations including nuclear magnetic resonance (NMR) spectroscopy