Learn More
Apexbio Technology LLC N1-Methylpseudouridine, 50mg CAS# 13860-38-3
SDP
Supplier:  Apexbio Technology LLC B834050
Encompass_Preferred
N1-Methylpseudouridine is a modified NTP that can be incorporated into mRNA via T7 RNA Polymerase. The addition of N1-methylpseudouridine can suppress immune response of the mRNA construct. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8261