Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N1,N1-diethyl-1-furan-2-yl-ethane-1,2-diamine

Catalog No. CDS007785 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS007785 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS007785 Supplier Sigma Aldrich Supplier No. CDS007785250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethyl orthobenzoate reacts with P-dimethylaminophosphonic acid bis(1-methylhydrazide) to yield 1,2,5,6-tetrahydro-1,5-dimethyl-6-(N,N-dimethylamino)-6-phenyl-1,2,4,5,6-tetrazaphosphorine-6-oxide.Trimethyl orthobenzoate was used in the preparation of 5-phenyl-4,6-dipyrrin and 2,3-orthoester derivatives of ethyl-1-thio-α-L-rhamnopyranoside.

Specifications

Molecular Formula C10H18N2O
Linear Formula C10H18N2O
MDL Number MFCD07643207
Quantity 50 g
Molecular Weight (g/mol) 182.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.