Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N1,N12-Diacetylspermine dihydrochloride | 77928-71-3 | 99.0% | 359.34 | 10 MG
SDP

Catalog No. 5000437035
Encompass_Preferred

N1,N12-Diacetylspermine dihydrochloride is a diacetylated derivative of Spermine. Its upregulation has been linked to the incidence of cancer, making it a potential biomarker for cancer detection.

  • Diacetylated derivative of spermine
  • Linked to cancer incidence, serving as a potential biomarker for detection
  • High purity of 99.0%
  • Supplied as a white to off-white solid
  • Recommended storage at -20°C in sealed, moisture-free conditions
  • Soluble in water at 100 mg/mL, requiring ultrasonication for dissolution
  • Intended for research use only

Catalog No. 50-004-37035 Supplier Medchemexpress LLC Supplier No. HY113374A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N1,N12-Diacetylspermine dihydrochloride is a diacetylated derivative of Spermine. Its upregulation has been linked to the incidence of cancer, making it a potential biomarker for cancer detection.

  • Diacetylated derivative of spermine
  • Linked to cancer incidence, serving as a potential biomarker for detection
  • High purity of 99.0%
  • Supplied as a white to off-white solid
  • Recommended storage at -20°C in sealed, moisture-free conditions
  • Soluble in water at 100 mg/mL, requiring ultrasonication for dissolution
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.