Learn More
Medchemexpress LLC N1,N12-Diacetylspermine dihydrochloride | 77928-71-3 | 99.0% | 359.34 | 100 MG
SDP
Catalog No. 5000438650
Encompass_Preferred
N1,N12-Diacetylspermine dihydrochloride is a diacetylated derivative of Spermine. Its upregulation has been linked to the incidence of cancer, suggesting its potential as a biomarker for cancer detection. This product is for research use only and not sold to patients.
  • Endogenous metabolite
  • Potential biomarker for cancer detection
Catalog No. 50-004-38650 Supplier Medchemexpress LLC Supplier No. HY113374A100MG
N1,N12-Diacetylspermine dihydrochloride is a diacetylated derivative of Spermine. Its upregulation has been linked to the incidence of cancer, suggesting its potential as a biomarker for cancer detection. This product is for research use only and not sold to patients.
  • Endogenous metabolite
  • Potential biomarker for cancer detection