Learn More
Medchemexpress LLC N1,N3,N5-tris(4-dodecylhexadecyl)benzene-1,3,5-tricarboxamide | 2922283-38-1 | 1385.4 g/mol | C93H177N3O3 | 5 MG
SDP
Catalog No. 5000221442
Encompass_Preferred
N1,N3,N5-Tris(4-dodecylhexadecyl)benzene-1,3,5-tricarboxamide is an ionizable lipid-like analogue intended for research use in nucleic acid delivery. It is primarily used in formulation and lipid nanoparticle (LNP) studies to evaluate delivery of mRNA, siRNA, and CRISPR/Cas components in preclinical research settings.
  • Ionizable lipid-like structure suitable for mRNA and nucleic acid delivery.
  • Designed for incorporation into lipid nanoparticle formulations.
  • Intended for laboratory research and formulation development.
  • Off-white to pale oil appearance, facilitating handling in solution.
  • Stable under recommended storage: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Not classified as a hazardous substance or mixture per supplier safety data.
Catalog No. 50-002-21442 Supplier Medchemexpress LLC Supplier No. HY1457985MG
N1,N3,N5-Tris(4-dodecylhexadecyl)benzene-1,3,5-tricarboxamide is an ionizable lipid-like analogue intended for research use in nucleic acid delivery. It is primarily used in formulation and lipid nanoparticle (LNP) studies to evaluate delivery of mRNA, siRNA, and CRISPR/Cas components in preclinical research settings.
  • Ionizable lipid-like structure suitable for mRNA and nucleic acid delivery.
  • Designed for incorporation into lipid nanoparticle formulations.
  • Intended for laboratory research and formulation development.
  • Off-white to pale oil appearance, facilitating handling in solution.
  • Stable under recommended storage: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Not classified as a hazardous substance or mixture per supplier safety data.