Learn More
Medchemexpress LLC N1,N3,N5-tris(4-dodecylhexadecyl)benzene-1,3,5-tricarboxamide | 2922283-38-1 | >98.0% | C93H177N3O3 | 10MG
SDP
Catalog No. 5000258444
Encompass_Preferred
N1,N3,N5-Tris(4-dodecylhexadecyl)benzene-1,3,5-tricarboxamide is an ionizable lipid-like compound used for research on lipid nanoparticle (LNP) formulations and nucleic acid delivery. It is applied in formulation optimization and preclinical mRNA delivery studies, and is supplied as a high-molecular-weight solid with supporting documentation for laboratory use.
  • Ionizable lipid-like structure suitable for LNP formulation.
  • Supports mRNA delivery and nucleic acid encapsulation studies.
  • High molecular weight and long alkyl chains enhance membrane interaction.
  • Provided as a research-grade solid with documentation for reproducible use.
Catalog No. 50-002-58444 Supplier Medchemexpress LLC Supplier No. HY14579810MG
N1,N3,N5-Tris(4-dodecylhexadecyl)benzene-1,3,5-tricarboxamide is an ionizable lipid-like compound used for research on lipid nanoparticle (LNP) formulations and nucleic acid delivery. It is applied in formulation optimization and preclinical mRNA delivery studies, and is supplied as a high-molecular-weight solid with supporting documentation for laboratory use.
  • Ionizable lipid-like structure suitable for LNP formulation.
  • Supports mRNA delivery and nucleic acid encapsulation studies.
  • High molecular weight and long alkyl chains enhance membrane interaction.
  • Provided as a research-grade solid with documentation for reproducible use.