Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N1,N5-Bis-boc-spermidine | 68076-39-1 | 99.8% | 345.48 | 100 MG
SDP

Catalog No. 5000403510
Encompass_Preferred

N1,N5-Bis-Boc-spermidine is a linker that features an amino group along with two Boc-protected amino groups. This amino group is reactive with various substances such as carboxylic acids, activated NHS esters, and carbonyls like ketone and aldehyde. The Boc group can be deprotected when exposed to mild acidic conditions, resulting in the formation of a free amine.

  • Linker containing an amino group with two Boc-protected amino groups.
  • Amino group is reactive with carboxylic acids, activated NHS esters, and carbonyls (ketone, aldehyde).
  • Boc group can be deprotected under mild acidic conditions to form the free amine.
  • Soluble in DMSO at a concentration of at least 100 mg/mL (289.45 mM).
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light).
  • Purity is 99.78%.

Catalog No. 50-004-03510 Supplier Medchemexpress LLC Supplier No. HY42222100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N1,N5-Bis-Boc-spermidine is a linker that features an amino group along with two Boc-protected amino groups. This amino group is reactive with various substances such as carboxylic acids, activated NHS esters, and carbonyls like ketone and aldehyde. The Boc group can be deprotected when exposed to mild acidic conditions, resulting in the formation of a free amine.

  • Linker containing an amino group with two Boc-protected amino groups.
  • Amino group is reactive with carboxylic acids, activated NHS esters, and carbonyls (ketone, aldehyde).
  • Boc group can be deprotected under mild acidic conditions to form the free amine.
  • Soluble in DMSO at a concentration of at least 100 mg/mL (289.45 mM).
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light).
  • Purity is 99.78%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.