Learn More
Medchemexpress LLC N1,N5-Bis-boc-spermidine | 68076-39-1 | 99.8% | 345.48 | 500 MG
SDP
Catalog No. 5000403506
Encompass_Preferred
N1,N5-Bis-Boc-spermidine is a linker molecule characterized by an amino group and two Boc-protected amino groups. The amino group is reactive with various functional groups such as carboxylic acids, activated NHS esters, and carbonyls (ketone, aldehyde). The Boc protecting groups can be removed under mild acidic conditions to yield the free amine.
  • Reactive amino group for conjugation with carboxylic acids, activated NHS esters, and carbonyls.
  • Boc protecting groups can be deprotected under mild acidic conditions.
  • Suitable for laboratory centrifugation and storage.
Catalog No. 50-004-03506 Supplier Medchemexpress LLC Supplier No. HY42222500MG
N1,N5-Bis-Boc-spermidine is a linker molecule characterized by an amino group and two Boc-protected amino groups. The amino group is reactive with various functional groups such as carboxylic acids, activated NHS esters, and carbonyls (ketone, aldehyde). The Boc protecting groups can be removed under mild acidic conditions to yield the free amine.
  • Reactive amino group for conjugation with carboxylic acids, activated NHS esters, and carbonyls.
  • Boc protecting groups can be deprotected under mild acidic conditions.
  • Suitable for laboratory centrifugation and storage.