Learn More
Medchemexpress LLC HY-129912A 5mg Medchemexpress, N1,N8-Diacetylspermidine (hydrochloride) CAS:178244-42-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY129912A5MG
Encompass_Preferred
Medchemexpress, HY-129912A 5mg N1,N8-Diacetylspermidine (hydrochloride) CAS:178244-42-3 N1,N8-Diacetylspermidine hydrochloride is a polyamines in the human urine. N1,N8-Diacetylspermidine hydrochloride is useful as prognostic indicators after treatment and during follow-up examination of cancer patients[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0176