Learn More
TARGETMOL CHEMICALS INC N106 5MG
SDP
Catalog No. 502580547
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. N106 is an activator of the SUMO-activating enzyme E1 ligase and triggers intrinsic sumoylation of SERCA2a. N106 can be used in studies about heart failure. purity: 98%
Catalog No. 50-258-0547 Supplier TARGETMOL CHEMICALS INC Supplier No. T281245MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. N106 is an activator of the SUMO-activating enzyme E1 ligase and triggers intrinsic sumoylation of SERCA2a. N106 can be used in studies about heart failure. purity: 98%