Learn More
AIRGAS Airgas® Model N114G320 Brass High Delivery Pressure Single Stage Regulator With 1/4" FNPT Connection
Catalog No. NC3791242
Encompass
Designed for a wide range of pressure settings, these single-stage regulators are recommended for use in petroleum field applications, research laboratories for hydrogenation, catalytic reduction, accelerated age testing, calorimetric testing, component testing, and pressure testing applications. These regulators feature a rugged brass piston, which provides increased safety and reliability at high delivery pressures. A 25-micron inlet filter protects internal components.
Catalog No. NC3791242 Supplier AIRGAS Supplier No. Y11N114G320AG
Designed for a wide range of pressure settings, these single-stage regulators are recommended for use in petroleum field applications, research laboratories for hydrogenation, catalytic reduction, accelerated age testing, calorimetric testing, component testing, and pressure testing applications. These regulators feature a rugged brass piston, which provides increased safety and reliability at high delivery pressures. A 25-micron inlet filter protects internal components.