Learn More
Cayman Chemical n1177trIfluoroacetAT salt 10mg
SDP
Catalog No. 502944499
Encompass_Preferred
A dual peptide GCGR and GLP-1R agonist; induces cAMP production in BHK-21 cells expressing human GCGR or GLP-1R (EC50s = 1.83 and 0.92 pM, respectively); reduces body weight and cumulative food intake in a mouse model of diet-induced obesity from 3-5 nmol/kg; reduces body weight, hepatic fat accumulation and fibrosis, and plasma ALT levels in a mouse model of NASH induced by a high-fat, high-fructose, and high-cholesterol diet
Catalog No. 50-294-4499 Supplier Cayman Chemical Supplier No. 4133310 MG
A dual peptide GCGR and GLP-1R agonist; induces cAMP production in BHK-21 cells expressing human GCGR or GLP-1R (EC50s = 1.83 and 0.92 pM, respectively); reduces body weight and cumulative food intake in a mouse model of diet-induced obesity from 3-5 nmol/kg; reduces body weight, hepatic fat accumulation and fibrosis, and plasma ALT levels in a mouse model of NASH induced by a high-fat, high-fructose, and high-cholesterol diet