Learn More
Cayman Chemical N1-Acetylspermidine (hydrochloride), Purity: ≥98%, CAS: 34450-16-3, 10 mg
SDP
Catalog No. 502893628
Encompass_Preferred
A substrate for polyamine oxidase that is oxidized to 3-acetamidopropanal and putrescine, a polyamine involved in the division, differentiation, and membrane function of cells; used to examine the interaction between DNA and polyamines during the cleavage of phosphodiester bonds at apurinic/apyrimidinic sites in DNA
Catalog No. 50-289-3628 Supplier Cayman Chemical Supplier No. 900153510 MG
A substrate for polyamine oxidase that is oxidized to 3-acetamidopropanal and putrescine, a polyamine involved in the division, differentiation, and membrane function of cells; used to examine the interaction between DNA and polyamines during the cleavage of phosphodiester bonds at apurinic/apyrimidinic sites in DNA