Learn More
Cayman Chemical N1-Acetylspermine (hydrochloride), Purity: ≥95%, CAS: 77928-70-2, 50 mg
SDP
Catalog No. 502968267
Encompass_Preferred
A monoacetylated derivative of spermine (Item No. 18041), an endogenous polyamine synthesized from spermidine (Item No. 14918), that displays lower Km and higher Vmax values than spermine, making it a better substrate of polyamine oxidase than the non-acetylated polyamine
Catalog No. 50-296-8267 Supplier Cayman Chemical Supplier No. 1791950 MG
A monoacetylated derivative of spermine (Item No. 18041), an endogenous polyamine synthesized from spermidine (Item No. 14918), that displays lower Km and higher Vmax values than spermine, making it a better substrate of polyamine oxidase than the non-acetylated polyamine