Learn More
Medchemexpress LLC N2-{[1-ethyl-6-(4-methylphenoxy)-1H-benzimidazol-2-yl]methyl}-L-alanineamide | 1233229-75-8 | 99.4% | 352.43 g/mol | C20H24N4O2 | 5 MG
SDP
Catalog No. 5000221119
Encompass_Preferred
Bliretrigine is a research-stage sodium channel blocker supplied for laboratory studies of pain and neuronal function. It is provided in high-purity solid and solution formats for preclinical experiments and is intended for research use only, not for human therapeutic use.
  • Sodium channel blocker with reported analgesic activity.
  • High purity suitable for research applications (99.4%).
  • Available as powder and as a DMSO solution.
  • Stable when stored under recommended conditions for powder and solutions.
  • Intended for research use only; not for human use.
Catalog No. 50-002-21119 Supplier Medchemexpress LLC Supplier No. HY1455585MG
Bliretrigine is a research-stage sodium channel blocker supplied for laboratory studies of pain and neuronal function. It is provided in high-purity solid and solution formats for preclinical experiments and is intended for research use only, not for human therapeutic use.
  • Sodium channel blocker with reported analgesic activity.
  • High purity suitable for research applications (99.4%).
  • Available as powder and as a DMSO solution.
  • Stable when stored under recommended conditions for powder and solutions.
  • Intended for research use only; not for human use.