Learn More
Medchemexpress LLC N2-(1H-Indol-5-yl)-6-(pyridin-4-yl)pyrazine-2,3-diamine acetate | 98.4% | 362.39 | 25 MG
SDP
Catalog No. 5000438167
Encompass_Preferred
AKN-028 acetate is a novel tyrosine kinase (TK) inhibitor and a potent, orally active FMS-like receptor tyrosine kinase 3 (FLT3) inhibitor with an IC50 value of 6 nM. It inhibits FLT3 autophosphorylation, induces a dose-dependent cytotoxic response (mean IC50=1 μM), and induces apoptosis by activation of caspase 3. AKN-028 acetate can be used in research of acute myeloid leukemia (AML).
  • Potent, orally active FMS-like receptor tyrosine kinase 3 (FLT3) inhibitor with an IC50 of 6 nM.
  • Inhibits FLT3 autophosphorylation.
  • Induces dose-dependent cytotoxic response (mean IC50=1 μM).
  • Induces apoptosis by activation of caspase 3.
  • Inhibits growth of primary AML and MV4-11 cells in mice.
  • Useful in research of acute myeloid leukemia (AML).
Catalog No. 50-004-38167 Supplier Medchemexpress LLC Supplier No. 25MGHY118304B
AKN-028 acetate is a novel tyrosine kinase (TK) inhibitor and a potent, orally active FMS-like receptor tyrosine kinase 3 (FLT3) inhibitor with an IC50 value of 6 nM. It inhibits FLT3 autophosphorylation, induces a dose-dependent cytotoxic response (mean IC50=1 μM), and induces apoptosis by activation of caspase 3. AKN-028 acetate can be used in research of acute myeloid leukemia (AML).
  • Potent, orally active FMS-like receptor tyrosine kinase 3 (FLT3) inhibitor with an IC50 of 6 nM.
  • Inhibits FLT3 autophosphorylation.
  • Induces dose-dependent cytotoxic response (mean IC50=1 μM).
  • Induces apoptosis by activation of caspase 3.
  • Inhibits growth of primary AML and MV4-11 cells in mice.
  • Useful in research of acute myeloid leukemia (AML).