Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-((4-methoxyphenyl)diphenylmethyl)-L-lysine | 159857-60-0 | MFCD00270542 | 98.1% | 640.77 g/mol | C41H40N2O5 | 100 G
SDP

Catalog No. 5000210613
Encompass_Preferred

N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-((4-methoxyphenyl)diphenylmethyl)-L-lysine is an Fmoc- and Dpm-protected L-lysine derivative used as a protected amino acid building block for solid-phase peptide synthesis. It is supplied as a solid with high purity and is intended for research use only.

  • Fmoc and Dpm protection for alpha and epsilon amino groups
  • Used as a building block in peptide synthesis
  • Cas number 159857-60-0
  • Molecular formula C41H40N2O5; molecular weight 640.77 g/mol
  • Purity 98.07% (HPLC)
  • Appearance solid; available in 5 g, 10 g, 25 g, 100 g sizes
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month)

Catalog No. 50-002-10613 Supplier Medchemexpress LLC Supplier No. HYW048704100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-((4-methoxyphenyl)diphenylmethyl)-L-lysine is an Fmoc- and Dpm-protected L-lysine derivative used as a protected amino acid building block for solid-phase peptide synthesis. It is supplied as a solid with high purity and is intended for research use only.

  • Fmoc and Dpm protection for alpha and epsilon amino groups
  • Used as a building block in peptide synthesis
  • Cas number 159857-60-0
  • Molecular formula C41H40N2O5; molecular weight 640.77 g/mol
  • Purity 98.07% (HPLC)
  • Appearance solid; available in 5 g, 10 g, 25 g, 100 g sizes
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.