Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-((4-methoxyphenyl)diphenylmethyl)-L-lysine | 159857-60-0 | MFCD00270542 | 98.1% | 640.77 g/mol | C41H40N2O5 | 5 G
SDP

Catalog No. 5000210714
Encompass_Preferred

N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-((4-methoxyphenyl)diphenylmethyl)-L-lysine is an Fmoc- and Mmt-protected L-lysine derivative used as an amino acid building block in solid-phase peptide synthesis and related research applications.

  • Contains Fmoc protection on the alpha-amino group
  • Contains Mmt protection on the side-chain epsilon-amino group
  • High purity suitable for research use (>98%)
  • Molecular weight 640.77 g/mol
  • Supplied as a 5 g quantity for laboratory use

Catalog No. 50-002-10714 Supplier Medchemexpress LLC Supplier No. HYW0487045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-((4-methoxyphenyl)diphenylmethyl)-L-lysine is an Fmoc- and Mmt-protected L-lysine derivative used as an amino acid building block in solid-phase peptide synthesis and related research applications.

  • Contains Fmoc protection on the alpha-amino group
  • Contains Mmt protection on the side-chain epsilon-amino group
  • High purity suitable for research use (>98%)
  • Molecular weight 640.77 g/mol
  • Supplied as a 5 g quantity for laboratory use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.