Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-carbamimidoyl-L-lysine | 776277-76-0 | 99.3% | 410.47 g/mol | C22H26N4O4 | 25 G
SDP

Catalog No. 5000208616
Encompass_Preferred

N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-carbamimidoyl-L-lysine is an Fmoc-protected lysine derivative bearing a carbamimidoyl modification on the N6 side chain, used as an amidino-lysine building block for peptide synthesis and research applications.

  • Cas number: 776277-76-0.
  • Molecular formula: C22H26N4O4.
  • Molecular weight: 410.47 g/mol.
  • Purity: 99.3% (reported by manufacturer).
  • Appearance: white to off-white solid.
  • Typical use: used as a protected arginine-like building block in peptide synthesis and related research.

Catalog No. 50-002-08616 Supplier Medchemexpress LLC Supplier No. 25GHYW089229
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-carbamimidoyl-L-lysine is an Fmoc-protected lysine derivative bearing a carbamimidoyl modification on the N6 side chain, used as an amidino-lysine building block for peptide synthesis and research applications.

  • Cas number: 776277-76-0.
  • Molecular formula: C22H26N4O4.
  • Molecular weight: 410.47 g/mol.
  • Purity: 99.3% (reported by manufacturer).
  • Appearance: white to off-white solid.
  • Typical use: used as a protected arginine-like building block in peptide synthesis and related research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.