Learn More
Medchemexpress LLC N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-(tert-butoxycarbonyl)-N2-methyl-L-lysine | 197632-76-1 | MFCD04973182 | 99.5% | 482.57 g/mol | C27H34N2O6 | 5 G
SDP
Catalog No. 5000299795
Encompass_Preferred
N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-(tert-butoxycarbonyl)-N2-methyl-L-lysine is an Fmoc- and Boc-protected N-methyl-L-lysine derivative supplied as a solid for use as a protected amino acid building block in solid-phase peptide synthesis and related research applications.
  • Fmoc and Boc protection for orthogonal deprotection
  • High purity suitable for peptide synthesis
  • Molecular weight 482.57 g/mol and formula C27H34N2O6
  • Available in multiple gram-scale package sizes
  • Certificate of analysis available with LCMS assay for batch-specific purity
Catalog No. 50-002-99795 Supplier Medchemexpress LLC Supplier No. HYW0363335G
N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-(tert-butoxycarbonyl)-N2-methyl-L-lysine is an Fmoc- and Boc-protected N-methyl-L-lysine derivative supplied as a solid for use as a protected amino acid building block in solid-phase peptide synthesis and related research applications.
  • Fmoc and Boc protection for orthogonal deprotection
  • High purity suitable for peptide synthesis
  • Molecular weight 482.57 g/mol and formula C27H34N2O6
  • Available in multiple gram-scale package sizes
  • Certificate of analysis available with LCMS assay for batch-specific purity