Learn More
Cayman Chemical N2-Isobutyryl-2-deoxygunosI 5g
SDP
Catalog No. 502942237
Encompass_Preferred
A nucleoside derivative and synthetic intermediate; has been used in the synthesis of relacin and nucleoside derivatives with antiviral activity against HIV-1
Catalog No. 50-294-2237 Supplier Cayman Chemical Supplier No. 402845 G
A nucleoside derivative and synthetic intermediate; has been used in the synthesis of relacin and nucleoside derivatives with antiviral activity against HIV-1