Learn More
Medchemexpress LLC N2-Isobutyryl-2'-O-methylguanosine | 63264-29-9 | 99.97% | 367.36 | 1 ML
SDP
Catalog No. 5000400614
Encompass_Preferred
N2-Isobutyryl-2'-O-methylguanosine is a nucleic acid synthesis intermediate utilized in applications such as antisense oligonucleotides and mRNA modification. It functions as a key monomer for synthesizing 2'-O-methyl oligoribonucleotides, allowing the final product to form stable double strands with complementary RNA and resist degradation by nucleases. It is primarily used in molecular biology research.
  • Used for antisense oligonucleotides and mRNA modification.
  • Serves as a key monomer for synthesizing 2'-O-methyl oligoribonucleotides.
  • Enables stable double strand formation with complementary RNA.
  • Resists degradation by nucleases.
  • Used in molecular biology research and pre-mRNA splicing mechanisms studies.
Catalog No. 50-004-00614 Supplier Medchemexpress LLC Supplier No. HY2128610MM/1ML
N2-Isobutyryl-2'-O-methylguanosine is a nucleic acid synthesis intermediate utilized in applications such as antisense oligonucleotides and mRNA modification. It functions as a key monomer for synthesizing 2'-O-methyl oligoribonucleotides, allowing the final product to form stable double strands with complementary RNA and resist degradation by nucleases. It is primarily used in molecular biology research.
  • Used for antisense oligonucleotides and mRNA modification.
  • Serves as a key monomer for synthesizing 2'-O-methyl oligoribonucleotides.
  • Enables stable double strand formation with complementary RNA.
  • Resists degradation by nucleases.
  • Used in molecular biology research and pre-mRNA splicing mechanisms studies.